Learn More
Medchemexpress LLC Thalidomide-propargyl | 2098487-39-7 | 99.97% | 312.28 | 10 MG
SDP
Catalog No. 5000436856
Encompass_Preferred
Thalidomide-propargyl is a Thalidomide-based Cereblon ligand used in the recruitment of CRBN protein. It can be connected to a ligand for protein by a linker to form IMiD containing PROTACs. It is a click chemistry reagent containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Thalidomide-based Cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can form IMiD containing PROTACs when connected to a ligand for protein by a linker
  • Click chemistry reagent
  • Contains an Alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-004-36856 Supplier Medchemexpress LLC Supplier No. HY12645710MG
Thalidomide-propargyl is a Thalidomide-based Cereblon ligand used in the recruitment of CRBN protein. It can be connected to a ligand for protein by a linker to form IMiD containing PROTACs. It is a click chemistry reagent containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Thalidomide-based Cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can form IMiD containing PROTACs when connected to a ligand for protein by a linker
  • Click chemistry reagent
  • Contains an Alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups