Learn More
Medchemexpress LLC HY-126457 5mg Medchemexpress, Thalidomide-propargyl CAS:2098487-39-7 Purity:>98%
SDP
Catalog No. 501969729
Encompass_Preferred
Medchemexpress, HY-126457 5mg Thalidomide-propargyl CAS:2098487-39-7 Thalidomide-propargyl is the Thalidomide-based Cereblon ligand used in the recruitment of CRBN protein. Thalidomide-propargyl can be connected to the ligand for protein by a linker to form the IMiD containing PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9729 Supplier Medchemexpress LLC Supplier No. HY1264575MG
Medchemexpress, HY-126457 5mg Thalidomide-propargyl CAS:2098487-39-7 Thalidomide-propargyl is the Thalidomide-based Cereblon ligand used in the recruitment of CRBN protein. Thalidomide-propargyl can be connected to the ligand for protein by a linker to form the IMiD containing PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.