Learn More
Medchemexpress LLC Thalidomide-Propargyne-PEG2-COOH | 2797619-65-7 | 98.7% | 428.39 | 25 MG
SDP
Catalog No. 5000350124
Encompass_Preferred
Thalidomide-Propargyne-PEG2-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates the thalidomide based cereblon ligand and a linker used in PROTAC technology. It is a click chemistry reagent, containing an alkyne group, and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Synthesized E3 ligase ligand-linker conjugate.
  • Incorporates thalidomide based cereblon ligand.
  • Uses a linker in PROTAC technology.
  • Click chemistry reagent.
  • Contains an alkyne group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc).
Catalog No. 50-003-50124 Supplier Medchemexpress LLC Supplier No. 25MGHY138777
Thalidomide-Propargyne-PEG2-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates the thalidomide based cereblon ligand and a linker used in PROTAC technology. It is a click chemistry reagent, containing an alkyne group, and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Synthesized E3 ligase ligand-linker conjugate.
  • Incorporates thalidomide based cereblon ligand.
  • Uses a linker in PROTAC technology.
  • Click chemistry reagent.
  • Contains an alkyne group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc).