Learn More
Medchemexpress LLC Thalidomide-propargyne-PEG2-COOH | 2797619-65-7 | 98.7% | C21H20N2O8 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13877710MG
Encompass_Preferred
Thalidomide-Propargyne-PEG2-COOH is a thalidomide-derived E3 ligase ligand-linker conjugate designed for use in PROTAC development and bioconjugation workflows. It incorporates a CRBN-binding thalidomide core, a short PEG2 spacer, and a terminal propargyne for click chemistry; supplied as a solid with high reported purity and good DMSO solubility.
Catalog No. 50-002-60549