Learn More
Medchemexpress LLC Thalidomide-propargyne-PEG3-COOH | 2797649-54-6 | 96.3% | C23H24N2O9 | 10MG
SDP
Catalog No. 5000260683
Encompass_Preferred
Thalidomide-Propargyne-PEG3-COOH is a thalidomide-derived E3 ligase ligand-linker conjugate featuring a terminal propargyl alkyne and a PEG3 spacer terminating in a carboxylic acid. It is intended as a research reagent for chemical biology and PROTAC synthesis to enable cereblon recruitment and bioorthogonal conjugation in degradation studies.
  • Provides a cereblon-binding ligand for CRBN-mediated degradation
  • Contains an alkyne handle for copper-catalyzed azide-alkyne cycloaddition
  • PEG3 spacer improves solubility and linker flexibility
  • Terminal carboxylic acid enables amide coupling and conjugation
  • Supplied at research-grade purity suitable for synthesis
Catalog No. 50-002-60683 Supplier Medchemexpress LLC Supplier No. HY13877610MG
Thalidomide-Propargyne-PEG3-COOH is a thalidomide-derived E3 ligase ligand-linker conjugate featuring a terminal propargyl alkyne and a PEG3 spacer terminating in a carboxylic acid. It is intended as a research reagent for chemical biology and PROTAC synthesis to enable cereblon recruitment and bioorthogonal conjugation in degradation studies.
  • Provides a cereblon-binding ligand for CRBN-mediated degradation
  • Contains an alkyne handle for copper-catalyzed azide-alkyne cycloaddition
  • PEG3 spacer improves solubility and linker flexibility
  • Terminal carboxylic acid enables amide coupling and conjugation
  • Supplied at research-grade purity suitable for synthesis