Learn More
Medchemexpress LLC Thalidomide-propargyne-PEG3-COOH | 2797649-54-6 | MFCD34473293 | 98.0% | 472.45 g/mol | C23H24N2O9 | 5 MG
SDP
Catalog No. 5000223314
Encompass_Preferred
Thalidomide-propargyne-PEG3-COOH is a thalidomide-based E3 ligase ligand-linker conjugate with a propargyl (alkynyl) handle and a PEG3 linker terminating in a carboxylic acid. Designed as a building block for PROTAC assembly and related chemical biology work, it is compatible with click chemistry and standard amide-coupling reactions. CAS 2797649-54-6; formula C23H24N2O9; MW 472.45; typical purity ~98%.
  • Contains a propargyl (alkynyl) handle for CuAAC click chemistry.
  • Includes a PEG3 linker terminating in a carboxylic acid for conjugation.
  • Acts as a cereblon (CRBN) E3 ligase ligand for PROTAC design.
  • Suitable for amide coupling and linker attachment.
  • Supplied as a solid with high purity for research use.
Catalog No. 50-002-23314 Supplier Medchemexpress LLC Supplier No. HY1387765MG
Thalidomide-propargyne-PEG3-COOH is a thalidomide-based E3 ligase ligand-linker conjugate with a propargyl (alkynyl) handle and a PEG3 linker terminating in a carboxylic acid. Designed as a building block for PROTAC assembly and related chemical biology work, it is compatible with click chemistry and standard amide-coupling reactions. CAS 2797649-54-6; formula C23H24N2O9; MW 472.45; typical purity ~98%.
  • Contains a propargyl (alkynyl) handle for CuAAC click chemistry.
  • Includes a PEG3 linker terminating in a carboxylic acid for conjugation.
  • Acts as a cereblon (CRBN) E3 ligase ligand for PROTAC design.
  • Suitable for amide coupling and linker attachment.
  • Supplied as a solid with high purity for research use.