Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-pyrrolidine hydrochloride | 2616553-35-4 | 98.0% | 335.74 g·mol⁻¹ | C15H14ClN3O4 | 100 MG
SDP

Catalog No. 5000280571
Encompass_Preferred

Thalidomide-pyrrolidine hydrochloride is an E3 ligase ligand (compound 18) used as a building block in the synthesis of PROTACs. Provided as the hydrochloride salt at research-grade purity, it is supplied in milligram-scale packages and includes storage recommendations for long-term stability.

  • E3 ligase ligand suitable for PROTAC assembly.
  • Hydrochloride salt form for improved handling.
  • High purity: 98.0%.
  • Molecular weight: 335.74 g·mol⁻¹.
  • CAS number: 2616553-35-4.
  • Available in multiple package sizes including 100 mg.
  • Storage recommendations: in solvent -80°C for 2 years, -20°C for 1 year.

Catalog No. 50-002-80571 Supplier Medchemexpress LLC Supplier No. HYW248665A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-pyrrolidine hydrochloride is an E3 ligase ligand (compound 18) used as a building block in the synthesis of PROTACs. Provided as the hydrochloride salt at research-grade purity, it is supplied in milligram-scale packages and includes storage recommendations for long-term stability.

  • E3 ligase ligand suitable for PROTAC assembly.
  • Hydrochloride salt form for improved handling.
  • High purity: 98.0%.
  • Molecular weight: 335.74 g·mol⁻¹.
  • CAS number: 2616553-35-4.
  • Available in multiple package sizes including 100 mg.
  • Storage recommendations: in solvent -80°C for 2 years, -20°C for 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.