Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-pyrrolidine hydrochloride | 2616553-35-4 | 98.0% | 335.74 g/mol | C15H14ClN3O4 | 25 MG
SDP

Catalog No. 5000280701
Encompass_Preferred

Thalidomide-pyrrolidine hydrochloride is an E3 ligase ligand (compound 18) used as a cereblon (CRBN) ligand in the synthesis of PROTAC degraders and other bifunctional molecules. It is supplied as an off-white to gray solid with high purity for research use.

  • High purity: 98.0%.
  • Molecular weight: 335.74 g/mol.
  • Molecular formula: C15H14ClN3O4.
  • CAS number: 2616553-35-4.
  • Physical form: off-white to gray solid.
  • Storage: store dry and keep cool; in solution store at -80°C (2 years) or -20°C (1 year).
  • Applications: used as an E3 ligase ligand for PROTAC synthesis.

Catalog No. 50-002-80701 Supplier Medchemexpress LLC Supplier No. HYW248665A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-pyrrolidine hydrochloride is an E3 ligase ligand (compound 18) used as a cereblon (CRBN) ligand in the synthesis of PROTAC degraders and other bifunctional molecules. It is supplied as an off-white to gray solid with high purity for research use.

  • High purity: 98.0%.
  • Molecular weight: 335.74 g/mol.
  • Molecular formula: C15H14ClN3O4.
  • CAS number: 2616553-35-4.
  • Physical form: off-white to gray solid.
  • Storage: store dry and keep cool; in solution store at -80°C (2 years) or -20°C (1 year).
  • Applications: used as an E3 ligase ligand for PROTAC synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.