Learn More
ION BIOSCIENCES Thallium-free GIRK Potassium Channel Assay pIONeer kit - 2 plates
Catalog No. NC4043227
Encompass_Preferred
ION Biosciences' Thallium-free GIRK Potassium Channel Assay pIONeer kit is the first fluorescence-based potassium channel solution that directly measures potassium flux, enabling you to move away from thallium-containing products. This product features the potassium-sensitive dye, IPG-1 AM, our lowest affinity potassium indicator to measure changes in intracellular potassium concentration. This kit is optimized for GIRK (Kir3.x) screening, and can be used to identify activators and inhibitors of GIRK channels. 2 plate kit.
Catalog No. NC4043227 Supplier ION BIOSCIENCES Supplier No. 120002
ION Biosciences' Thallium-free GIRK Potassium Channel Assay pIONeer kit is the first fluorescence-based potassium channel solution that directly measures potassium flux, enabling you to move away from thallium-containing products. This product features the potassium-sensitive dye, IPG-1 AM, our lowest affinity potassium indicator to measure changes in intracellular potassium concentration. This kit is optimized for GIRK (Kir3.x) screening, and can be used to identify activators and inhibitors of GIRK channels. 2 plate kit.