Learn More
ION BIOSCIENCES Thallium-free hERG Potassium Channel Assay pIONeer kit - 2 plates
Catalog No. NC4043220
Encompass_Preferred
ION Biosciences' Thallium-free hERG Potassium Channel Assay pIONeer kit is the first fluorescence-based, potassium channel solution that directly measures potassium flux, enabling you to move away from thallium-containing products. This product features the potassium-sensitive dye, IPG-1 AM, our lowest affinity potassium indicator to measure changes in intracellular potassium concentration. This kit is optimized for hERG screening (Kv11.1), and can be used to identify activators and inhibitors of hERG channels. This direct-readout eliminates the need for surrogate ions or radioactive tracers and enables robust, high-throughput screening to identify hERG modulators in 96- and 384-well formats. 2 plate kit.
Catalog No. NC4043220 Supplier ION BIOSCIENCES Supplier No. 121002
ION Biosciences' Thallium-free hERG Potassium Channel Assay pIONeer kit is the first fluorescence-based, potassium channel solution that directly measures potassium flux, enabling you to move away from thallium-containing products. This product features the potassium-sensitive dye, IPG-1 AM, our lowest affinity potassium indicator to measure changes in intracellular potassium concentration. This kit is optimized for hERG screening (Kv11.1), and can be used to identify activators and inhibitors of hERG channels. This direct-readout eliminates the need for surrogate ions or radioactive tracers and enables robust, high-throughput screening to identify hERG modulators in 96- and 384-well formats. 2 plate kit.