Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Thallium(I) acetate

Catalog No. 503125950 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-5950 100 g
50-312-5946 25 g
50-312-5947 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-312-5950 Supplier Sigma Aldrich Supplier No. T8266100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Thallium (I) acetate can be prepared by dissolving thallium (III) oxide in acetic acid and acetic anhydride and stirring at 80-90oC. After filtration, the solution cooled and the product is recrystallized. A study reports that a large number of mycoplasmas and acholeplasmas can grow in its presence at concentration of 1 mg/ml.Commonly used in the laboratories as a selective element against gram-negative bacteria in selective media for the detection of mollicutes. It may be used in synthesizing thallium derivatives.

Specifications

CAS 563-68-8
Quantity 100 g
Molecular Formula C2H3O2Tl
Linear Formula CH3COOTl
MDL Number MFCD00013045
Synonym Acetic acid thallium(I) salt
Molecular Weight (g/mol) 263.43
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.