Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thallium(I) bromide, ultra dry, 99.998% (metals basis)

Catalog No. AA3572009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3572009 10 g
AA3572018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3572009 Supplier Thermo Scientific Chemicals Supplier No. 03572009
Add to Cart

CAS: 7789-40-4

Thallium(I) bromide is used in blue sensitive photodetectors. It is also used as a real-time x-ray image sensor. Further, it acts as a standard for elemental thallium. In addition to this, it is a compound semiconductor used in room temperature X- and gamma-ray detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-40-4
MDL Number MFCD00011273
Synonym Thallous bromide

Specifications

Melting Point ∼460°C
Physical Form Beads
Quantity 10 g
UN Number UN1707
Merck Index 14,9258
Solubility Information Insoluble in water.
Formula Weight 284.28
Percent Purity 99.998%
Packaging Ampule
Sensitivity Hygroscopic
Density 7.5 g/mL
Assay (metals basis)
Chemical Name or Material Thallium(I) bromide, Ultra dry
Show More Show Less
Hazard Category H300+H330-H373
Hazard Statement GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
EINECSNumber 232-163-0
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.