Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thallium(I) chloride, ultra dry, 99.999% (metals basis)

Catalog No. AA3572106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3572106 5 g
AA3572114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3572106 Supplier Thermo Scientific Chemicals Supplier No. 03572106
Add to Cart

CAS: 7791-12-0 | ClTl | 239.83 g/mol

It is used as a catalyst in chlorination's and in suntan lamps. The low solubility of TlCl is exploited in chemical synthesis: treatment of metal chloride complexes with TlPF6, gives the corresponding metal hexafluorophosphate derivative. It is used as a tracer in medical imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-12-0
Molecular Formula ClTl
Molecular Weight (g/mol) 239.83
MDL Number MFCD00011274
InChI Key GBECUEIQVRDUKB-UHFFFAOYSA-M
Synonym thallium chloride, thallium i chloride, thallous chloride, thallium monochloride, thallium 1+ chloride, tlcl, thallium chloride tlcl, thallium chloride van, rcra waste number u216, rcra waste no. u216
PubChem CID 24642
ChEBI CHEBI:37117
IUPAC Name chlorothallium
SMILES Cl[Tl]

Specifications

Melting Point 430°C
Physical Form Beads
Quantity 5 g
UN Number UN1707
Merck Index 14,9260
Solubility Information Slightly soluble in water. Insoluble in alcohol,and acetone.
Formula Weight 239.82
Percent Purity 99.999%
Packaging Ampule
Refractive Index 2.247
Sensitivity Air Sensitive
Density 7.0 g/mL
Assay (metals basis)
Chemical Name or Material Thallium(I) chloride, Ultra dry
Show More Show Less
Hazard Category H300+H330-H373
Hazard Statement GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
EINECSNumber 232-241-4
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.