Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thallium(I) iodide, ultra dry, 99.999% (metals basis)

Catalog No. p-4904550
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3572209 10 g
AA3572218 50 g
AA3572230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3572209 Supplier Thermo Scientific Chemicals Supplier No. 03572209
Only null left
Add to Cart

CAS: 7790-30-9 | ITl | 331.28 g/mol

Thallium(I) iodide is used in mercury lamp to enhance its performance. It is mainly used for underwater lighting, since the blue green light produced has least absorption in water. Scintillator made by using iodide salt of thallium, sodium and cesium is utilized for radiation scintillation detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-30-9
Molecular Formula ITl
Molecular Weight (g/mol) 331.28
MDL Number MFCD00011279
InChI Key CMJCEVKJYRZMIA-UHFFFAOYSA-M
Synonym thallium iii iodide, thallium iodide tli2, thallium iodide ous, thallium i iodide, anhydrous, ampuled under argon trace metals basis 10g, tli
IUPAC Name λ¹-thallanylium iodide
SMILES [I-].[Tl+]

Specifications

Boiling Point 824°C
Melting Point 440°C
Color Yellow
Physical Form Beads
Quantity 10 g
Assay Percent Range 99.999% metal basis
Solubility Information Insoluble in water.
Formula Weight 331.27
Odor Odorless
Sensitivity Light sensitive
Density 7.1
Chemical Name or Material Thallium(I) iodide, ultra dry
Show More Show Less
Hazard Category H300+H330-H373
Precautionary Statement P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 232-199-7
RTECSNumber XG5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.