Learn More
Thallium(I) neodecanoate, (Tl 52.5-55.5%), Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Physical Form | Solid |
| Quantity | 50 g |
| UN Number | UN1707 |
| Formula Weight | 375.61 |
| Concentration or Composition (by Analyte or Components) | Tl 52.5 to 55.5% |
| Chemical Name or Material | Thallium(I) neodecanoate |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H300-H330-H373 Fatal if swallowed. Fatal if inhaled. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | GHS P Statement P260-P301+P310-P304+P340-P320-P330-P405-P501a Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Specific treatment is urgent (see label). Rinse mouth. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.