Learn More
Strem, An Ascensus Company CAS# 1314-32-5. 10g. Thallium(III) oxide (99.99%-Tl) PURATREM. MFCD00011276
SDP
Supplier:  Strem, An Ascensus Company 81810010G
Encompass
CAS# 1314-32-5. 10g. Thallium(III) oxide (99.99%-Tl) PURATREM. MFCD00011276. Molecular Weight: 456.74. Molecular Formula: Tl2O3. Color/form: black pwdr. Strem# 81-8100. http://www.strem.com/catalog/v/81-8100/
Catalog No. 50-901-13498