Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thallium Iodide (Ous), 99.6%, MP Biomedicals

Catalog No. ICN211769 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN211769 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN211769 Supplier MP Biomedicals, Inc Supplier No. 0521176910

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7790-30-9
Molecular Formula ITl
Molecular Weight (g/mol) 331.28
MDL Number MFCD00011279
InChI Key CMJCEVKJYRZMIA-UHFFFAOYSA-M
Synonym thallium iii iodide, thallium iodide tli2, thallium iodide ous, thallium i iodide, anhydrous, ampuled under argon trace metals basis 10g, tli
IUPAC Name λ¹-thallanylium iodide
SMILES [I-].[Tl+]

Specifications

Quantity 10 g
Formula Weight 331.31
Chemical Name or Material Thallium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.