Learn More
Medchemexpress LLC THAM-d11 (for molecular biology) | 202656-13-1 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYD0227ES100MG
Encompass_Preferred
Trometamol(Tris)-d11 for molecular biology is a deuterium labeled compound. It is for research use only and serves as a valuable tool in various analytical applications. This compound can be utilized as a tracer and an internal standard for quantitative analysis. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-10231