Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC THAM hydrochloride | 1185-53-1 | 99.67% | 500 G
SDP

Supplier:  Medchemexpress LLC HYD0227J500G

Encompass_Preferred

THAM hydrochloride (≥99%, for cell culture) is a biochemical reagent intended for research use only.

  • Biochemical reagent for cell culture
  • Molecular weight: 157.60
  • Formula: C4H12ClNO3
  • Appearance: Solid
  • Color: White to off-white
  • Shipping at room temperature in continental US
  • Storage at -20°C, sealed, away from moisture
  • Soluble in H2O at ≥ 100 mg/mL
  • Soluble in DMSO at 35 mg/mL

Catalog No. 50-004-10284


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.