Learn More
Medchemexpress LLC THAM hydrochloride (Tris HCl) | 1185-53-1 | 98.0% | 500 G
SDP
Catalog No. 5000410153
Encompass_Preferred
THAM hydrochloride (Tris HCl) is a biologically inert amino alcohol of low toxicity, which buffers carbon dioxide and acids in vitro and in vivo. It is an effective amine compound for pH control in the physiological range.
  • Biologically inert amino alcohol of low toxicity
  • Buffers carbon dioxide and acids in vitro and in vivo
  • Effective amine compound for pH control in the physiological range
Catalog No. 50-004-10153 Supplier Medchemexpress LLC Supplier No. HYD0227A500G
THAM hydrochloride (Tris HCl) is a biologically inert amino alcohol of low toxicity, which buffers carbon dioxide and acids in vitro and in vivo. It is an effective amine compound for pH control in the physiological range.
  • Biologically inert amino alcohol of low toxicity
  • Buffers carbon dioxide and acids in vitro and in vivo
  • Effective amine compound for pH control in the physiological range