Learn More
Aviva Systems Biology Corp THAP6 Antibody - middle region (ARP52758_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52758_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against THAP6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human THAP6. Alternate names: MGC30052ApplicationWB. Species reactivity: Dog, Horse, Human, Pig, Rat
Catalog No. 50-285-762