Learn More
AdipoGen Thapsigargin (high purity),Chemical. CAS: 67526-95-8
SDP
Supplier:  AdipoGen AGCN20003M001
Encompass_Preferred
Thapsigargin (high purity), AdipoGen Life Sciences. Chemical. CAS: 67526-95-8. Formula: C34H50O12. MW: 650.8. Isolated from Thapsia garganica. Intracellular Ca2+ signaling probe. Specific and sensitive inhibitor of SERCA. Non-TPA/PMA tumor promoter. Histamine release inducer. Apoptosis inducer. Mitochondrial dysfunction inducer. NOS modulator. Angiogenesis inhibitor. Stimulator of arachidonic acid metabolism in macrophages. Autophagy inducer. TRAIL sensitizer.
Catalog No. 50-464-293