Learn More
Medchemexpress LLC HY-13433 1mg Medchemexpress, Thapsigargin CAS:67526-95-8 Purity:>98%
SDP
Catalog No. 502028994
Encompass_Preferred
Medchemexpress, HY-13433 1mg Thapsigargin CAS:67526-95-8 Thapsigargin is an inhibitor of microsomal Ca2+-ATPase. Thapsigargin is a well characterized Endoplasmic Reticulum (ER) stress inducing agent[2][3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8994 Supplier Medchemexpress LLC Supplier No. HY134331MG
Medchemexpress, HY-13433 1mg Thapsigargin CAS:67526-95-8 Thapsigargin is an inhibitor of microsomal Ca2+-ATPase. Thapsigargin is a well characterized Endoplasmic Reticulum (ER) stress inducing agent[2][3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.