Learn More
Selleck Chemical LLC Thapsigargin 1mg
SDP
Catalog No. 502173321
Encompass_Preferred
Thapsigargin is a potent, non-competitive inhibitor ofthe sarco/endoplasmic reticulum Ca2+ ATPase (SERCA) with IC50 of 0.353 nM or 0.448 nM for the carbachol-evoked [Ca2+]i-transients with or without a KCl-prestimulation. Thapsigargin induces cellapoptosis. Thapsigargin is extracted from a plant, Thapsia garganica. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3321 Supplier Selleck Chemical LLC Supplier No. S78951MG
Thapsigargin is a potent, non-competitive inhibitor ofthe sarco/endoplasmic reticulum Ca2+ ATPase (SERCA) with IC50 of 0.353 nM or 0.448 nM for the carbachol-evoked [Ca2+]i-transients with or without a KCl-prestimulation. Thapsigargin induces cellapoptosis. Thapsigargin is extracted from a plant, Thapsia garganica. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.