Learn More
Enzo Life Sciences Thapsigargin, (1mg), CAS Number: 67526-95-8
SDP
Catalog No. NC9006970
Encompass_Preferred
ATPase inhibitor. Formula: C34H50O12. MW: 650.8. CAS Number: 67526-95-8. MI: 14: 9272. Purity: ≥95% (HPLC). Appearance: Colorless wax or white solid. Solubility: Soluble in DMSO (20mg/ml), 100% ethanol (20mg/ml) or acetone. Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at -20°C. Long Term Storage: -20°C
Catalog No. NC9006970 Supplier Enzo Life Sciences Supplier No. BMLPE1800001
ATPase inhibitor. Formula: C34H50O12. MW: 650.8. CAS Number: 67526-95-8. MI: 14: 9272. Purity: ≥95% (HPLC). Appearance: Colorless wax or white solid. Solubility: Soluble in DMSO (20mg/ml), 100% ethanol (20mg/ml) or acetone. Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at -20°C. Long Term Storage: -20°C