Learn More
AdipoGen Thapsigargin
SDP
Catalog No. 502559956
Encompass_Preferred
Chemical. CAS 67526-95-8. Formula C34H50O12. Molecular Weight 650.75. Thapsigargin is a non-competitive, cell permeable inhibitor of calcium transport by SERCAs IC50 values are cell type-dependent and range from ~2-80 nM. Inhibition of SERCAs leads to an increase in intracellular calcium, which has been linked to cell activation, release of histamine from mast cells, and increased proliferation of certain types of cancer cells. Thapsigargin shows anti-inflammatory and anti-cancer effects.
Catalog No. 50-255-9956 Supplier AdipoGen Supplier No. CDXT0284M005
Chemical. CAS 67526-95-8. Formula C34H50O12. Molecular Weight 650.75. Thapsigargin is a non-competitive, cell permeable inhibitor of calcium transport by SERCAs IC50 values are cell type-dependent and range from ~2-80 nM. Inhibition of SERCAs leads to an increase in intracellular calcium, which has been linked to cell activation, release of histamine from mast cells, and increased proliferation of certain types of cancer cells. Thapsigargin shows anti-inflammatory and anti-cancer effects.