Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thapsigargin, 99% (TLC), 97.6% (HPLC), MP Biomedicals™

Catalog No. ICN15899905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15899905 5 mg
ICN15899901 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15899905 Supplier MP Biomedicals, Inc Supplier No. 0215899905
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67526-95-8
Molecular Formula C34H50O12
Molecular Weight (g/mol) 650.76
MDL Number MFCD00083511
InChI Key IXFPJGBNCFXKPI-FSIHEZPISA-N
Synonym thapsigargin, unii-z96bq26rzd, z96bq26rzd, chembl96926, octanoic acid 3s-3alpha, 3abeta, 4alpha, 6beta, 6abeta, 7beta, 8alpha z , 9balpha-6-acetyloxy-2,3,-3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy-azuleno 4,5-b furan-7-yl ester, octanoic acid, 6-acetyloxy-2,3,3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy azuleno 4,5-b furan-7-yl ester, 3s-3alpha,3abeta,4alpha,6beta,6abeta,7beta,8alpha z ,9balpha, tg1, --thapsigargin, octanoic acid 3s-3alpha, 3abeta, 4alpha, 6beta,6abeta, 7beta, 8alpha z , 9balpha-6-acetyloxy-2,3,-3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy-azuleno 4,5-b furan-7-yl ester, octanoic acid 3s-3alpha, 3abeta, 4alpha,6beta, 6abeta, 7beta, 8alpha z , 9balpha-6-acetyloxy-2,3,-3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy-azuleno 4,5-b furan-7-yl ester
PubChem CID 126969181
IUPAC Name (3S,3aR,4S,6S,6aR,7S,8S,9bS)-6-(acetyloxy)-4-(butanoyloxy)-3,3a-dihydroxy-3,6,9-trimethyl-8-{[(2Z)-2-methylbut-2-enoyl]oxy}-2-oxo-2H,3H,3aH,4H,5H,6H,6aH,7H,8H,9bH-azuleno[4,5-b]furan-7-yl octanoate
SMILES CCCCCCCC(=O)O[C@@H]1[C@@H](OC(=O)C(\C)=C/C)C(C)=C2[C@@H]3OC(=O)[C@@](C)(O)[C@@]3(O)[C@H](C[C@](C)(OC(C)=O)[C@@H]12)OC(=O)CCC

Specifications

Color White
Assay Percent Range 99% (TLC), 97.6% (HPLC)
Quantity 5 mg
Formula Weight 650.76
Physical Form Crystalline Powder
Chemical Name or Material Thapsigargin, For TLC and HPLC analysis
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.