Learn More
Abcam Thapsigargin, Ca2+-ATPase inhibitor, 1MG
Catalog No. 502584635
Encompass_Preferred
MW 650.8 Da, Purity >98%. A potent, cell-permeable Ca2+-ATPase inhibitor. Releases Ca2+ by inhibiting endoplasmic reticular Ca2+-ATPase (IC50 = 4-13 nM). Both tumorogenic and apoptotic actions reported.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4635 Supplier Abcam Supplier No. AB1202861MG
Add to Cart
Add to Cart
MW 650.8 Da, Purity >98%. A potent, cell-permeable Ca2+-ATPase inhibitor. Releases Ca2+ by inhibiting endoplasmic reticular Ca2+-ATPase (IC50 = 4-13 nM). Both tumorogenic and apoptotic actions reported.
The product is subject to the following: Abcam Restricted Use Statement