Learn More
Eurofins Discoverx AssayComplete Thawing Reagent T5, 100. (92-4105TR)
Catalog No. 501999863
Encompass
AssayComplete Thawing Reagent T5 is an optimized thawing reagent for revival and recovery of Eurofins DiscoverX cell lines. This reagent ensures optimal post-thaw viability, recovery and assay performance upon thawing. This product is provided as a bottle of 100 mL. Please refer to cell line specific datasheet to determine the recommended thawing reagent. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.
Catalog No. 50-199-9863 Supplier Eurofins Discoverx Supplier No. 924105TR
AssayComplete Thawing Reagent T5 is an optimized thawing reagent for revival and recovery of Eurofins DiscoverX cell lines. This reagent ensures optimal post-thaw viability, recovery and assay performance upon thawing. This product is provided as a bottle of 100 mL. Please refer to cell line specific datasheet to determine the recommended thawing reagent. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.