Learn More
Eurofins Discoverx AssayComplete Thawing Reagent T8, 100. (92-4108TR)
Catalog No. 501999864
Encompass
AssayComplete Thawing Reagent T8 is an optimized thawing reagent for revival and recovery of Eurofins DiscoverX cell lines. This reagent ensures optimal post-thaw viability, recovery and assay performance upon thawing. This product is provided as a bottle of 100 mL. Please refer to cell line specific datasheet to determine the recommended thawing reagent. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.
Catalog No. 50-199-9864 Supplier Eurofins Discoverx Supplier No. 924108TR
AssayComplete Thawing Reagent T8 is an optimized thawing reagent for revival and recovery of Eurofins DiscoverX cell lines. This reagent ensures optimal post-thaw viability, recovery and assay performance upon thawing. This product is provided as a bottle of 100 mL. Please refer to cell line specific datasheet to determine the recommended thawing reagent. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.