Learn More
STEMCELL Technologies ThawSTAR CFT2 Automated Thawing System
SDP
Catalog No. NC2016823
Encompass
Increase confidence in your cell thawing workflow and ensure sample sterility and consistent thawing performance by using the ThawSTAR CFT2 Automated Thawing System. With a standardized thawing process that replaces manual, water bath-based thawing, ThawSTAR eliminates the risk of contamination and delivers controlled thawing profiles. Using this convenient and compact system, you can thaw your cells directly in the biosafety cabinet. This system is compatible with a variety of 1.8 - 2.0 mL vials.Simply insert a frozen cryogenic vial and retrieve it once the device alerts you at the end of the thaw cycle, after approximately 2.5 minutes. The ThawSTAR CFT2 Automated Thawing System can be leveraged early in the R&D phase and scaled into commercial manufacturing.Additional products available for purchase include ThawSTAR CFT2 Transporter, ThawSTAR CFT2 IOPQ Ki and ThawSTAR CFT2 Confirmation Vials.
Catalog No. NC2016823 Supplier STEMCELL Technologies Supplier No. 1000650
Increase confidence in your cell thawing workflow and ensure sample sterility and consistent thawing performance by using the ThawSTAR CFT2 Automated Thawing System. With a standardized thawing process that replaces manual, water bath-based thawing, ThawSTAR eliminates the risk of contamination and delivers controlled thawing profiles. Using this convenient and compact system, you can thaw your cells directly in the biosafety cabinet. This system is compatible with a variety of 1.8 - 2.0 mL vials.Simply insert a frozen cryogenic vial and retrieve it once the device alerts you at the end of the thaw cycle, after approximately 2.5 minutes. The ThawSTAR CFT2 Automated Thawing System can be leveraged early in the R&D phase and scaled into commercial manufacturing.Additional products available for purchase include ThawSTAR CFT2 Transporter, ThawSTAR CFT2 IOPQ Ki and ThawSTAR CFT2 Confirmation Vials.