Learn More
STEMCELL Technologies ThawSTAR CFT2 IOPQ Kit, Size: 1 Kit
SDP
Catalog No. NC2016077
Encompass
The ThawSTAR CFT2 IOPQ Kit includes installation, operational, and performance qualification documentation and testing tool accessories to facilitate functional testing of the ThawSTAR CFT2 Automated Thawing System (/products/thawstar-cft2-automated-thawing-system.html). Providing a water-free and standardized cell thawing process, the ThawSTAR CFT2 Automated Thawing System comes complete with a ThawSTAR CFT2 Transporter, two ThawSTAR CFT2 Confirmation Vials, and ThawSTAR CFT2 Power Supply and Adaptors. Use the ThawSTAR CFT2 IOPQ Kit for a step-by-step IOPQ protocol and summary report templates that are based upon GxP industry standards and the GAMP 5 methodology.For more information on protocols for cell thawing using ThawSTAR, please explore the Technical Manual (/products/thawstar-cft2-iopq-kit.html#section-protocols-and-documentation).
Catalog No. NC2016077 Supplier STEMCELL Technologies Supplier No. 1000730
The ThawSTAR CFT2 IOPQ Kit includes installation, operational, and performance qualification documentation and testing tool accessories to facilitate functional testing of the ThawSTAR CFT2 Automated Thawing System (/products/thawstar-cft2-automated-thawing-system.html). Providing a water-free and standardized cell thawing process, the ThawSTAR CFT2 Automated Thawing System comes complete with a ThawSTAR CFT2 Transporter, two ThawSTAR CFT2 Confirmation Vials, and ThawSTAR CFT2 Power Supply and Adaptors. Use the ThawSTAR CFT2 IOPQ Kit for a step-by-step IOPQ protocol and summary report templates that are based upon GxP industry standards and the GAMP 5 methodology.For more information on protocols for cell thawing using ThawSTAR, please explore the Technical Manual (/products/thawstar-cft2-iopq-kit.html#section-protocols-and-documentation).