Learn More
STEMCELL Technologies ThawSTAR CFT2 Transporter, Size: 1 Unit
SDP
Catalog No. 502390353
Encompass
Portable solution for the handling and transport of frozen vials from liquid nitrogen or -80C freezer
Catalog No. 50-239-0353 Supplier STEMCELL Technologies Supplier No. 1000642
Portable solution for the handling and transport of frozen vials from liquid nitrogen or -80C freezer