Learn More
AdipoGen Thaxtomin C
SDP
Supplier:  AdipoGen BVT0391M001
Encompass_Preferred
Chemical. CAS 140111-05-3. Formula C21H20N4O4. MW 392.4. Isolated from Streptomyces bottropensis Gö-Dra 17. Diketopiperazine antibiotic. Biosynthetic precursor of Thaxtomin A Prod.No. BVT-0206 http//www.adipogen.com/bvt-0206/thaxtomin-a.html . Phytotoxin. Plant cell necrosis inducer. Induces common scab disease of potato.
Catalog No. 50-256-1956