Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Thayer Martin Modified Agar with VCNT
SDP

Catalog No. 23001732 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Species:
Neisseria gonorrhoeae
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
23-001-732 Neisseria gonorrhoeae
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-732 Supplier Hardy Diagnostics™ Supplier No. E30
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the selective isolation of Neisseria gonorrhoeae.

  • Contains vancomycin, colistin, nystatin, and trimethoprim antimicrobials

Specifications

Appearance Opaque
Color Brown
Content And Storage 2°C to 8°C
Country of Origin United States
Additive Colistin, Trimethoprim Lactate, Vancomycin, Nystatin, KoEnzyme Enrichments
Detects Neisseria gonorrhoeae
Form Prepared Medium
Format Plate
Product Type Agar Plate
Target Organism Class Gram-negative Microorganisms
Test Type Qualitative QC
Fill Volume 18 mL
Formulation Proteose Peptone No. 3 - 15.0gm, Hemoglobin, Bovine - 10.0gm, Sodium Chloride - 5.0gm, Dipotassium Phosphate - 4.0gm, Monopotassium Phosphate - 1.0gm, Corn Starch - 1.0gm, Colistin - 7.5mg, Trimethoprim Lactate - 5.0mg, Vancomycin - 3.0mg, Nystatin - 1,250U, KoEnzyme Enrichments - 10.0ml, Agar - 12.0gm
For Use With (Application) Selective Isolation
Incubation Conditions 35°C
Incubation Time 24 to 48 hr.
No. per Pack 10
Packaging Type Plates, Bag
Quantity 10/Pk.
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Neisseria gonorrhoeae
Protein Source Proteose Peptone No. 3, Hemoglobin, Bovine
Source Bovine
Manufacturing Quality ISO 13485
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.