Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7056MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: THBD/1782. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: THP1 Cells. Placenta, Angiosarcoma, Bladder or Cervical Carcinoma. Cellular Staining: Cell Surface ;Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human Thrombomodulin protein fragment (aa69-194). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 100kDa.Chromosome: 20p11.2. Gene Symbol: THBD. Gene ID: 7056. Ensemble: ENSG00000178726. Uniprot: NP_000352.
Catalog No. 50-188-5219