Learn More
Medchemexpress LLC THBS1 Human HEK293 100ug
SDP
Catalog No. 5000196921
Encompass_Preferred
Recombinant human thrombospondin-1 (THBS1) produced in HEK293 cells and supplied as a lyophilized, C-terminal His-tagged protein. Suitable for biochemical assays and cell-based studies that require full-length, glycosylated THBS1; lot-specific certificates of analysis provide formulation and purity details.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • C-terminal His tag enables purification and detection.
  • Includes amino acids N19-P1170 of human THBS1.
  • Supplied lyophilized for stability and long-term storage.
  • Purity greater than 85% by reducing SDS-PAGE.
  • Lot-specific COA documents formulation, molecular weight, and analytical data.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O.
Catalog No. 50-001-96921 Supplier Medchemexpress LLC Supplier No. HYP70725100UG
Recombinant human thrombospondin-1 (THBS1) produced in HEK293 cells and supplied as a lyophilized, C-terminal His-tagged protein. Suitable for biochemical assays and cell-based studies that require full-length, glycosylated THBS1; lot-specific certificates of analysis provide formulation and purity details.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • C-terminal His tag enables purification and detection.
  • Includes amino acids N19-P1170 of human THBS1.
  • Supplied lyophilized for stability and long-term storage.
  • Purity greater than 85% by reducing SDS-PAGE.
  • Lot-specific COA documents formulation, molecular weight, and analytical data.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O.