Learn More
GA International THC-Standardized Cannabis Symbol (Canada) Labels 0.75" x 0.75" / 19.1mm x 19.1mm Rectangle White on Red From -40°C to +93°C (-40°F to +199.4°F)
Catalog No. 5000571186
Encompass_Preferred
Pre-printed, red and black on a white background, THC content warning labels. These permanent labels are made of a durable thermoplastic film, and resist abrasion, scratching, as well as spraying and wiping with alcohols, detergents, and surface sanitizing wipes. Ideal for labeling a wide range of cannabis products (THC concentration of 10 micrograms/gram and higher) packaging containers, such as plastic bags, bottles, and cardboard boxes. These standardized cannabis symbol labels are also UV resistant, and tearproof, able to withstand a wide range of temperatures ( -40ºC to +93ºC / -40ºF to +200ºF), perfect for use in any environment, and storage conditions.
Catalog No. 50-005-71186 Supplier GA International Supplier No. WLA0181P
Pre-printed, red and black on a white background, THC content warning labels. These permanent labels are made of a durable thermoplastic film, and resist abrasion, scratching, as well as spraying and wiping with alcohols, detergents, and surface sanitizing wipes. Ideal for labeling a wide range of cannabis products (THC concentration of 10 micrograms/gram and higher) packaging containers, such as plastic bags, bottles, and cardboard boxes. These standardized cannabis symbol labels are also UV resistant, and tearproof, able to withstand a wide range of temperatures ( -40ºC to +93ºC / -40ºF to +200ºF), perfect for use in any environment, and storage conditions.