Learn More
GA International THC-Standardized Cannabis Symbol (Canada) Labels 1" x 1" / 25.4mm x 25.4mm Rectangle White on Red From -40°C to +93°C (-40°F to +199.4°F)
Catalog No. 5000571449
Encompass_Preferred
Pre-printed, red and black on a white background, THC content warning labels. These permanent labels are made of a durable thermoplastic film, and resist abrasion, scratching, as well as spraying and wiping with alcohols, detergents, and surface sanitizing wipes. Ideal for labeling a wide range of cannabis products (THC concentration of 10 micrograms/gram and higher) packaging containers, such as plastic bags, bottles, and cardboard boxes. These standardized cannabis symbol labels are also UV resistant, and tearproof, able to withstand a wide range of temperatures (-40ºC to +93ºC / -40ºF to +200ºF), perfect for use in any environment, and storage conditions.
Catalog No. 50-005-71449 Supplier GA International Supplier No. WLA0171P
Pre-printed, red and black on a white background, THC content warning labels. These permanent labels are made of a durable thermoplastic film, and resist abrasion, scratching, as well as spraying and wiping with alcohols, detergents, and surface sanitizing wipes. Ideal for labeling a wide range of cannabis products (THC concentration of 10 micrograms/gram and higher) packaging containers, such as plastic bags, bottles, and cardboard boxes. These standardized cannabis symbol labels are also UV resistant, and tearproof, able to withstand a wide range of temperatures (-40ºC to +93ºC / -40ºF to +200ºF), perfect for use in any environment, and storage conditions.