Learn More
Electron Microscopy Sciences Thick Carbon Conductive Tabs, 12 mm
SDP
Catalog No. 5025108
Encompass_Preferred
Stiff smooth surface tabs have 200 um conductive carbonate base, coated 30 μ thick on each side with conductive adhesive. 260 μ thick. Not as conductive and sticky as the Standard carbon Conductive Tabs.
Diameter: 12 mm
Catalog No. 50-251-08 Supplier Electron Microscopy Sciences Supplier No. 7782412
Stiff smooth surface tabs have 200 um conductive carbonate base, coated 30 μ thick on each side with conductive adhesive. 260 μ thick. Not as conductive and sticky as the Standard carbon Conductive Tabs.
Diameter: 12 mm