Learn More
Bachem a-Synuclein (61-95) (human) Trifluoroacetate
SDP
Supplier:  Bachem 4026207.0500
Encompass_Preferred
a-Synuclein (61-95) (human) Trifluoroacetate, H-Glu-Gln-Val-Thr-Asn-Val-Gly-Gly-Ala-Val-Val-Thr-Gly-Val-Thr-Ala-Val-Ala-Gln-Lys-Thr-Val-Glu-Gly-Ala-Gly-Ser-Ile-Ala-Ala-Ala-Thr-Gly-Phe-Val-OH, 154040-19-4, C141H235N39O49, Mr 3260.65, 0.5mg. Non-Ab Component of Alzheimer's Disease Amyloid, NAC, Non-b-Amyloid Component of Alzheimer's Disease (NAC). The 35 amino acid peptide NAC (a-synuclein (61-95)) has originally been isolated from the insoluble core of Alzheimer's disease (AD) amyloid plaque. The sequence corresponds to a fragment of a 140 amino acid precursor known as NACP or a-synuclein and was found to form amyloid fibrils via a nucleation-dependent polymerization mechanism. Accumulation of NAC aggregates in the synapse might be responsible for the neurodegeneration in AD and in the prion diseases.
Catalog No. 50-194-6650