Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PELTOR™ CH-3 Listen Only Hearing Protector

Catalog No. 19072249 Shop All 3M Products
Change view
Click to view available options
Model:
HT52B-112
HT52P3E-112
Style:
Hard Hat
Neckband
Noise Reduction Rating (NRR):
25 dB
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Style Noise Reduction Rating (NRR)
19-072-249 HT52B-112 Neckband 25 dB
19-072-263 HT52P3E-112 Hard Hat 25 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-072-249 Supplier 3M™ Supplier No. 7100203298
Only null left
Add to Cart
Add to Cart

PELTOR™ CH-3 Listen Only Hearing Protector helps workers stay protected in noisy environments while enabling them to hear incoming audio from handheld devices.

  • Hearing protection solution with clear audio reception in noisy environments
  • FLX2 connection jack for easy and reliable connection to a wide assortment of handheld devices
  • Lasting comfort for long duration wear times
  • Designed to withstand harsh environmental elements
  • Audio speakers are optimized for speech frequencies and meet the efficiency demands of many of today's digital devices
  • Bright yellow color for enhanced visibility

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Color Bright Yellow
ANSI S3.1974 Yes
Cushion Color Black
For Use With (Application) Manufacturing
High Visibility Yes
Model HT52B-112
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 25 dB
Product Type CH-3 Listen Only
Dielectric No
Style Neckband
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.