Learn More
Perkin Elmer US LLC T-Clip Adapter for 50 mm Transmission-Reflectance Sphere
Catalog No. 502016332
Encompass_Preferred
The 50 mm Transmission-Reflectance Sphere is used for measuring diffuse reflectance (specular component included SCI), total transmittance or hemispherical transmittance, and performing color analysis. It has a spectral range of 2301100 nm (UV and visible). Recommended for solid color analysis.
Catalog No. 50-201-6332 Supplier Perkin Elmer US LLC Supplier No. N4101500
The 50 mm Transmission-Reflectance Sphere is used for measuring diffuse reflectance (specular component included SCI), total transmittance or hemispherical transmittance, and performing color analysis. It has a spectral range of 2301100 nm (UV and visible). Recommended for solid color analysis.