Learn More
Bachem D-Biotinyl-OSu
SDP
Supplier:  Bachem 4014225.1000
Encompass_Preferred
D-Biotinyl-OSu, 35013-72-0, C14H19N3O5S, Mr 341.39, 1g. BNHS, Biotin-NHS. The activated biotin derivative Biotin-OSu is used for labeling the amino groups of amino acids, peptides, and proteins. When conducting the biotinylation at pH 6.5, N-terminal amino groups can be selectively tagged in the presence of lysine.
Catalog No. 50-194-7400