Learn More
Bachem Tau Peptide (306-336) (Repeat 3 Domain) Trifluoroacetate
SDP
Supplier:  Bachem 4095430.1000
Encompass_Preferred
Tau Peptide (306-336) (Repeat 3 Domain) Trifluoroacetate, H-Val-Gln-Ile-Val-Tyr-Lys-Pro-Val-Asp-Leu-Ser-Lys-Val-Thr-Ser-Lys-Cys-Gly-Ser-Leu-Gly-Asn-Ile-His-His-Lys-Pro-Gly-Gly-Gly-Gln-OH, 330456-26-3, C143H236N42O42S, Mr 3247.77, 1mg. The amphipathic helical structure of the third fragment (306-336) in the four-repeat microtubule-binding domain of the water-soluble tau protein could be responsible for the formation of the neuropathological filament.
Catalog No. 50-194-6640