Learn More
Bachem Amyloid Beta Protein (1-43)
SDP
Supplier:  Bachem 4018358.1000
Encompass_Preferred
Amyloid Beta Protein (1-43), H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-Thr-OH, 134500-80-4, C207H318N56O62S, Mr 4615.21, 1mg. b-Amyloid (1-43). The amyloid b-protein is a 39- to 43-amino acid polypeptide that is the primary constituent of senile plaques and cerebrovascular deposits in Alzheimer's disease and Down's syndrome. Additionally it acts as an inhibitor of the ubiquitin-dependent protein degradation in vitro.
Catalog No. 50-194-6594