Learn More
SOMNI Scientific™ The Animal Temperature Heating Pad
SDP
Catalog No. 04777177 Shop All Somni Scientific Products
Change view
Click to view available options
For Use With (Application):
Research Animal Inhalant Anesthesia
Catalog No. For Use With (Application)
04-777-177 Research Animal Inhalant Anesthesia
1 options
Catalog No. 04-777-177 Supplier SOMNI Scientific™ Supplier No. HT15003
Add to Cart
Add to Cart
Ultra-thin, low noise heating pad that offers uniform heating to maintain the animal’s body temperature. Low input voltage reduces electric noise generated from the heating current
  • Offers uniform heating to maintain the animal’s body temperature
  • Low input voltage reduces electric noise generated from the heating current
  • Ultra-thin
  • 140 x 80 mm
  • Low noise 
  • Range 0-50°C
  • Available with international adapter
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
Temperature (Metric) Range 0°C to 50°C