Learn More
Bachem Lactoferricin B25 Trifluoroacetate
SDP
Supplier:  Bachem 4087382.0005
Encompass_Preferred
Lactoferricin B25 Trifluoroacetate, H-Phe-Lys-Cys-Arg-Arg-Trp-Gln-Trp-Arg-Met-Lys-Lys-Leu-Gly-Ala-Pro-Ser-Ile-Thr-Cys-Val-Arg-Arg-Ala-Phe-OH, 146897-68-9, C141H224N46O29S3, Mr 3123.82, 5mg. LFcinB 25, Lactoferricin B (17-41) (bovine). The antimicrobial peptide lactoferricin B25 is a fragment of cow's milk protein lactoferrin. The peptide exhibits potent dose-dependent and time-dependent anticancer activity against human stomach cancer cell cultures via apoptosis and autophagy. LFcinB25 is expected to yield novel anticancer drugs.
Catalog No. 50-194-6676