Learn More
Bachem (Gly22)-Amyloid Beta Protein (1-42)
SDP
Supplier:  Bachem 4035371.1000
Encompass_Preferred
(Gly22)-Amyloid Beta Protein (1-42) , H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Gly-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH, 1802086-23-2, C200H307N55O58S, Mr 4442.04, 1mg. Amyloid b-Protein (1-42) Arctic Mutation, Amyloid b-Protein (1-42) E22G. The arctic mutant of amyloid b peptide 1-42, in which Glu22 is substituted by Gly, is distinctly more amyloidogenic than the wild-type Ab 1-42.
Catalog No. 50-194-6611