Learn More
Perkin Elmer US LLC Peripump Tubing Pharmx 2X2 mm 1x3.2 mm
Catalog No. 502016464
Encompass_Preferred
The autosampler has a 3-channel peristaltic pump that accepts below listed tubes when running inorganic acids. 2 2 mm ID Pharmx, Black Tabs, Rinse 1 3.2mm ID Pharmx, White Tabs, Waste
Catalog No. 50-201-6464 Supplier Perkin Elmer US LLC Supplier No. N0831030
The autosampler has a 3-channel peristaltic pump that accepts below listed tubes when running inorganic acids. 2 2 mm ID Pharmx, Black Tabs, Rinse 1 3.2mm ID Pharmx, White Tabs, Waste